Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Antibody (PAb 240), PerCP, Novus Biologicals™
SDP

Catalog No. NB200103PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200103PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200103PCP Supplier Novus Biologicals Supplier No. NB200103PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p53 Monoclonal specifically detects p53 in Human, Mouse, Rat, Yeast, Xenopus (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen p53
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PAb 240
Conjugate PerCP
Dilution Flow Cytometry, Flow (Intracellular)
Gene Alias Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Gene Symbols TP53
Host Species Mouse
Immunogen Gel-purified p53-beta-galactosidase fusion protein containing murine p53 from aa 14-389 (derived from pSV53C cDNA clone).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7157
Test Specificity This monoclonal recognizes both mutant forms and wild-type human p53 under denaturing conditions. It does not recognize nondenatured wild type p53. In nondenaturing conditions (IP) it only recognizes mutant p53.
Target Species Human, Mouse, Rat, Yeast, Xenopus (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.