Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Antibody (V5P4H6*B3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20231A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20231A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20231A94 Supplier Novus Biologicals Supplier No. NBP250231AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p53 Monoclonal specifically detects p53 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen p53
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V5P4H6*B3
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Gene Symbols TP53
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KLMVKDEGRS (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7157.0
Target Species Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.