Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 DINP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | p53 DINP1 |
---|---|
Dilution | Western Blot 1:1000-1:5000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056627T
![]() |
Novus Biologicals
NB10056627SS |
0.05 mL |
Each for $206.50
|
|
|||||
NB10056627
![]() |
Novus Biologicals
NB10056627 |
0.2 mL |
Each for $426.00
|
|
|||||
Description
p53 DINP1 Polyclonal specifically detects p53 DINP1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
p53 DINP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, DNA Double Strand Break Repair | |
Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
DKFZp434M1317, FLJ22139, p53DINP1, P53DINP1p53-dependent damage-inducible nuclear protein 1, p53-inducible p53DINP1, SIPStress-induced protein, Teap, TP53DINP1, TP53INP1A, TP53INP1B, tumor protein p53 inducible nuclear protein 1, tumor protein p53-inducible nuclear protein 1 | |
TP53INP1 | |
IgG | |
Unpurified |
Western Blot 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
Q96A56 | |
94241 | |
This antibody was generated by immunizing rabbit with synthetic peptides of human p53DINP1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title