Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 DINP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056627 0.2 mL
NB10056627T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056627 Supplier Novus Biologicals Supplier No. NB10056627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p53 DINP1 Polyclonal specifically detects p53 DINP1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen p53 DINP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000
Formulation Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Accession No. Q96A56
Gene Alias DKFZp434M1317, FLJ22139, p53DINP1, P53DINP1p53-dependent damage-inducible nuclear protein 1, p53-inducible p53DINP1, SIPStress-induced protein, Teap, TP53DINP1, TP53INP1A, TP53INP1B, tumor protein p53 inducible nuclear protein 1, tumor protein p53-inducible nuclear protein 1
Gene Symbols TP53INP1
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbit with synthetic peptides of human p53DINP1.
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Double Strand Break Repair
Primary or Secondary Primary
Gene ID (Entrez) 94241
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.