Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ p53 (Mutant), Mouse, Unlabeled, Clone: PAb240,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ OP29200UG
Description
Recognizes the ∽53kDa mutant p53 protein under non-denaturing conditions by immunoprecipitation, immunofluorescence, and flow cytometry. Recognizes both mutant and wild-type p53 by immunoblotting and paraffin sections under denaturing conditions.
Specifications
p53 (Mutant) | |
Monoclonal | |
Please refer to vial label for lot-specific concentration | |
In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5, ≤0.1% Sodium Azide. | |
a recombinant protein consisting of amino acids 14-389 of p53 fused to β-galactosidase | |
RUO | |
Bovine | |
Liquid |
Gel Supershift, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PAb240 | |
Unconjugated | |
Mouse | |
200 μg | |
Primary | |
2°-8°C | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction