Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ p53 (Mutant), Mouse, Unlabeled, Clone: PAb240,
Mouse Monoclonal Antibody
$525.84 - $575.12
Specifications
Antigen | p53 (Mutant) |
---|---|
Clone | PAb240 |
Concentration | Please refer to vial label for lot-specific concentration |
Applications | Gel Supershift, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Recognizes the ∽53kDa mutant p53 protein under non-denaturing conditions by immunoprecipitation, immunofluorescence, and flow cytometry. Recognizes both mutant and wild-type p53 by immunoblotting and paraffin sections under denaturing conditions.Specifications
p53 (Mutant) | |
Please refer to vial label for lot-specific concentration | |
Monoclonal | |
Liquid | |
RUO | |
In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5, ≤0.1% Sodium Azide. | |
IgG1 | |
2°-8°C |
PAb240 | |
Gel Supershift, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Bovine | |
a recombinant protein consisting of amino acids 14-389 of p53 fused to β-galactosidase | |
Primary |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title