Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Acetylated p53 (K382) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | p53 |
---|---|
Clone | 2589A |
Dilution | Western Blot 4 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13552SP
![]() |
R&D Systems
MAB13552SP |
25 μg |
Each for $200.50
|
|
|||||
MAB13552100
![]() |
R&D Systems
MAB13552100 |
100 μg |
Each for $467.50
|
|
|||||
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
p53 | |
Western Blot 4 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
P04637 | |
7157 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2589A | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Synthetic peptide containing acetylated human p53 (K382), Accession # P04637 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title