Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 Rat anti-Mouse, Unconjugated, Clone: POE316A/E9, MilliporeSigma™
Rat Monoclonal Antibody
$189.29 - $481.52
Specifications
Antigen | p53 |
---|---|
Clone | POE316A/E9 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE18082MI
|
MilliporeSigma
MABE180825UG |
25 μg |
Each for $189.29
|
|
|||||
MABE18081MI
|
MilliporeSigma
MABE1808100UG |
100 μg |
Each for $481.52
|
|
|||||
Description
p53 Monoclonal antibody specifically detects p53 in Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
p53 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Mouse | |
Cellular tumor antigen p53;Tumor suppressor p53 | |
GST/His-tagged full-length recombinant mouse p53 protein. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
43.46 kDa calculated. |
POE316A/E9 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_035770.2 | |
Tp53, P53, Trp53 | |
IgG2a κ | |
Protein G Purified | |
Clone POE316A/E9 is a rat monoclonal antibody that detects murine p53. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title