Learn More
Description
Specifications
Specifications
| Antigen | p53R2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 74 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.17.4.1, MGC102856, MGC42116, MTDPS8A, MTDPS8B, p53-inducible ribonucleotide reductase small subunit 2 homolog, p53-inducible ribonucleotide reductase small subunit 2-like protein, p53R2, P53R2DKFZp686M05248, PEOA5, ribonucleoside-diphosphate reductase subunit M2 B, ribonucleotide reductase M2 B (TP53 inducible), TP53-inducible ribonucleotide reductase M2 B |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human p53R2 (Q7LG56-1; Met1-Phe351). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
