Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p62/SQSTM1 Antibody (2533B), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB027867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB027867 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB027867 Supplier Novus Biologicals Supplier No. NBP292995AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

p62/SQSTM1 Monoclonal specifically detects p62/SQSTM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen p62/SQSTM1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 2533B
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Host Species Rabbit
Immunogen Partial recombinant human p62/SQSTM1 protein produced in E. coli (amino acids 368-440) [UniProt Q13501]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8878
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.