Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p62/SQSTM1 Antibody (864807), HRP, Novus Biologicals™
SDP

Catalog No. NB373746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373746 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373746 Supplier Novus Biologicals Supplier No. FAB8028H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p62/SQSTM1 Monoclonal antibody specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Gene Knock-Out

Specifications

Antigen p62/SQSTM1
Applications Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Gene Knock-Out
Classification Monoclonal
Clone 864807
Conjugate HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Knockout Validated
Formulation PBS
Gene Alias A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Host Species Mouse
Immunogen E. coli-derived recombinant human p62/SQSTM1, Asp368-Leu440, Accession # Q13501
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8878
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.