Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p62/SQSTM1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP142821SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142821SS 0.025 mL
NBP142821 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP142821SS Supplier Novus Biologicals Supplier No. NBP142821SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen p62/SQSTM1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50
Formulation PBS, 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. Q13501, Q64337, O08623
Gene Alias A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Gene Symbols SQSTM1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 350-400). [Swiss-Prot Q13501]
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8878
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.