Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p62/SQSTM1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 6 publications

$192.47 - $519.67

Specifications

Antigen p62/SQSTM1
Concentration 1 mg/mL
Dilution Western Blot 0.5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP142821SS
SDP
View Documents
Novus Biologicals
NBP142821SS
0.025 mL
Each for $192.47
Only null left
Add to Cart
 
NBP142821
SDP
View Documents
Novus Biologicals
NBP142821
0.1 mL
Each for $519.67
Only null left
Add to Cart
 
Description

Description

p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

p62/SQSTM1
Western Blot 0.5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50
Polyclonal
Rabbit
Autophagy, Signal Transduction
PBS, 30% Glycerol with 0.1% Sodium Azide
A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
SQSTM1
IgG
Affinity Purified
47 kDa
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Rat
Q13501, Q64337, O08623
8878
A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 350-400). [Swiss-Prot Q13501]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.