Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p66 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056643SS
Description
p66 alpha Polyclonal specifically detects p66 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
p66 alpha | |
Polyclonal | |
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:50 | |
Q8CHY6 | |
GATAD2A | |
This antibody was developed against a synthetic peptide corresponding to aa 137-150 of mouse p66a. It cross-reacts with human p66a (AAH19178.1). | |
Unpurified | |
RUO | |
54815 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antisera with 0.05% Sodium Azide | |
FLJ20085, FLJ21017, GATA zinc finger domain containing 2A, GATA zinc finger domain-containing protein 2A, Hp66alpha, p66 alpha, p66alpha, transcriptional repressor p66-alpha | |
Rabbit | |
68 kDa | |
0.05 mL | |
Primary | |
Human, Mouse, Rat | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction