Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p66 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | p66 alpha |
---|---|
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:50 |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056643T
![]() |
Novus Biologicals
NB10056643SS |
0.05 mL |
Each for $181.90
|
|
|||||
NB10056643
![]() |
Novus Biologicals
NB10056643 |
0.2 mL |
Each for $402.00
|
|
|||||
Description
p66 alpha Polyclonal specifically detects p66 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
p66 alpha | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8CHY6 | |
54815 | |
This antibody was developed against a synthetic peptide corresponding to aa 137-150 of mouse p66a. It cross-reacts with human p66a (AAH19178.1). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:50 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with 0.05% Sodium Azide | |
FLJ20085, FLJ21017, GATA zinc finger domain containing 2A, GATA zinc finger domain-containing protein 2A, Hp66alpha, p66 alpha, p66alpha, transcriptional repressor p66-alpha | |
GATAD2A | |
IgG | |
Unpurified | |
68 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title