Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p67phox/NOXA2 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB322217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322217 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322217 Supplier Novus Biologicals Supplier No. NBP297097PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p67phox/NOXA2 Polyclonal antibody specifically detects p67phox/NOXA2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen p67phox/NOXA2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias autosomal 2, NADPH oxidase activator 2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2FLJ93058, p67phox, p67-phox
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human p67phox/NOXA2 (Accession#: AAH01606.1; Met1-Val526)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4688
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.