Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p67phox/NOXA2 Rabbit anti-Mouse, Rat, Clone: 7J7N3, Novus Biologicals™
SDP

Catalog No. NB167605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167605 100 μg
NB167606 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167605 Supplier Novus Biologicals Supplier No. NBP316257100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

p67phox/NOXA2 Monoclonal antibody specifically detects p67phox/NOXA2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen p67phox/NOXA2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7J7N3
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias autosomal 2, NADPH oxidase activator 2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2FLJ93058, p67phox, p67-phox
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human p67phox/NOXA2 (P19878).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4688
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.