Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p70 S6 Kinase/S6K Antibody (OTI1G4), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273209UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273209UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273209UV Supplier Novus Biologicals Supplier No. NBP273209UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p70 S6 Kinase/S6K Monoclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen p70 S6 Kinase/S6K
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1G4
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A
Gene Symbols RPS6KB1
Host Species Mouse
Immunogen Full length recombinant protein of human p70 S6 Kinase/S6K (NP_003152) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Autophagy, Cancer, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6198.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.