Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p70 S6 Kinase/S6K, p Thr389/412 Antibody, Novus Biologicals™
SDP

Catalog No. NB600104901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600104901 0.1 mg
NB413992 50 μg
NB018203 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB600104901 Supplier Novus Biologicals Supplier No. NB60010490.1MG

Rabbit Polyclonal Antibody has been used in 6 publications

p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen p70 S6 Kinase/S6K (p Thr389)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UBS0
Gene Alias EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A
Gene Symbols RPS6KB1
Host Species Rabbit
Immunogen Synthetic phospho-peptide surrounding amino acid Thr389 of human p70 S6 kinase
Purification Method Affinity Purified
Quantity 0.1 mg
Research Discipline Cancer, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6198
Test Specificity This detects p70 S6 Kinase only when phosphorylated at Thr389.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.