Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p70 S6 Kinase/S6K, p Thr389/412 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$438.34
Specifications
| Antigen | p70 S6 Kinase/S6K (p Thr389) |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB413992
![]() |
Novus Biologicals
NB600104950UG |
50 μg |
Each for $438.34
|
|
|||||
NB018203
![]() |
Novus Biologicals
NB60010490.025MG |
0.025 mg | N/A | N/A | N/A | ||||
NB600104901
![]() |
Novus Biologicals
NB60010490.1MG |
0.1 mg | N/A | N/A | N/A | ||||
Description
p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| p70 S6 Kinase/S6K (p Thr389) | |
| Western Blot | |
| Unconjugated | |
| Cancer, mTOR Pathway, Phospho Specific | |
| PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal | |
| EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A | |
| RPS6KB1 | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Rat, Human, Mouse, Rabbit | |
| Q9UBS0 | |
| 6198 | |
| Synthetic phospho-peptide surrounding amino acid Thr389 of human p70 S6 kinase | |
| Primary | |
| This detects p70 S6 Kinase only when phosphorylated at Thr389. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title