Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Antibody (5B1288), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10056674R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056674R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056674R Supplier Novus Biologicals Supplier No. NB10056674R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p73 Monoclonal specifically detects p73 in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen p73
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5B1288
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. O15350
Gene Alias p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Gene Symbols TP73
Host Species Mouse
Immunogen This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418).
Molecular Weight of Antigen 73 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7161
Test Specificity It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. NB100-56674 has been shown to recognize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.