Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Antibody (5B1288), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339490 Supplier Novus Biologicals Supplier No. NB10056674MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p73 Monoclonal antibody specifically detects p73 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)

Specifications

Antigen p73
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 5B1288
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Host Species Mouse
Immunogen This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.