Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Antibody (5B429), PE, Novus Biologicals™
SDP

Catalog No. NBP224737PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224737PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224737PE Supplier Novus Biologicals Supplier No. NBP224737PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p73 Monoclonal specifically detects p73 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen p73
Applications Flow Cytometry
Classification Monoclonal
Clone 5B429
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. O15350
Gene Alias p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Gene Symbols TP73
Host Species Mouse
Immunogen This antibody was raised against full-length human p73. It reacts with an epitope that is located on the N-terminal region of human p73. It does not cross react with p53.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7161
Test Specificity p73 antibody (clone 5B429) recognizes all of the TAp73 isoforms i.e. isoforms with an intact NH2-terminal transactivation domain (Thottassery et el, 2006, PMID 16505115). Sayan et al 2005 (PMID 15781249) showed that p73 IMG-246/ NBP2-24737 reacted with all tested TAp73 isoforms but not deltaNp73 whereas p73 antibody clone 5B1288 (NB100-56674/ IMG-259A ) showed reactivity against all p73 isoforms including delta Np73. Caspase-cleaved p73 fragments, including 55 kDa and 45 kDa, have been described in literature (Sayan et al, 2008).
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.