Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB292018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292018 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292018 Supplier Novus Biologicals Supplier No. NB100420F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p73 Polyclonal antibody specifically detects p73 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen p73
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region between residues 1 and 62 of human tumor protein p73 using the numbering given in Swiss-Prot entry O15350 (GeneID 7161).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.