Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Antibody (P73/2531), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB384622 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384622 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384622 Supplier Novus Biologicals Supplier No. NBP308760IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p73 Monoclonal antibody specifically detects p73 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen p73
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone P73/2531
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Host Species Mouse
Immunogen Recombinant full-length human p73 protein (Uniprot: O15350)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.