Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ P7C3

Click to view available options
Quantity:
5 mg
25 mg
Description
P7C3 is a NAMPT activator. Proneurogenic and neuroprotective. Protects newborn neurons in the dentate gyrus by mitigating cell death. Also enhances learning and memory in aged rats. Orally available and brain penetrant. Also revents doxorubicin-mediated toxicity in osteosarcoma cells and restores intracellular NAD levels.
Specifications
Specifications
Chemical Name or Material | 3,6-Dibromo-α-[(phenylamino)methyl]-9H-carbazole-9-ethanol |
CAS | 301353-96-8 |
Assay Percent Range | >99% |
Quantity | 25 mg |
Molecular Formula | C21H18Br2N2O |
Formula Weight | Observed MW: 474.19 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction