Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p97/VCP Antibody, Novus Biologicals™
SDP

Catalog No. NB1001557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001557 Supplier Novus Biologicals Supplier No. NB1001557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

p97/VCP Polyclonal specifically detects p97/VCP in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen p97/VCP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:15000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 1-4 ug/mg of lysate, Immunohistochemistry-Paraffin 1:200- 1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_009057.1
Gene Alias 15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog
Gene Symbols VCP
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Valosin-Containing Protein using the numbering given in entry NP_009057.1 (GeneID 7415).
Molecular Weight of Antigen 89 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7415
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.