Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB358669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358669 Supplier Novus Biologicals Supplier No. NBP176239MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1 Polyclonal antibody specifically detects PA1 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen PA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias chromosome 16 open reading frame 53, FLJ22459, GAS, glutamate-rich coactivator interacting with SRC1/NCOA1, MGC4606, PA1, PAXIP1-associated protein 1, PTIP-associated 1 protein, PTIP-associated protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human GAS. The immunogen is located within the last 50 amino acids of GAS. Amino Acid Squence: LDSEDPSPASPPLRS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79447
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.