Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA28 Activator alpha Subunit/PSME1 Antibody (016), HRP, Novus Biologicals™
SDP

Catalog No. NB038682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038682 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038682 Supplier Novus Biologicals Supplier No. NBP290298H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PA28 Activator alpha Subunit/PSME1 Monoclonal specifically detects PA28 Activator alpha Subunit/PSME1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PA28 Activator alpha Subunit/PSME1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 16
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias 11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PA28 Activator alpha Subunit/PSME1 (Q06323; Met1-Tyr249).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5720
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.