Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA28 Activator alpha Subunit/PSME1 Antibody (12H3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB161230 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161230 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161230 Supplier Novus Biologicals Supplier No. NBP259434FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PA28 Activator alpha Subunit/PSME1 Monoclonal antibody specifically detects PA28 Activator alpha Subunit/PSME1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen PA28 Activator alpha Subunit/PSME1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 12H3
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias 11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha
Host Species Mouse
Immunogen Recombinant human PA28 Activator alpha Subunit/PSME1 (1-249aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 5720
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.