Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA28 Activator beta Subunit/PSME2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB065698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065698 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065698 Supplier Novus Biologicals Supplier No. NBP300350AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA28 Activator beta Subunit/PSME2 Polyclonal specifically detects PA28 Activator beta Subunit/PSME2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PA28 Activator beta Subunit/PSME2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PA28 Activator beta Subunit/PSME2 (Q9UL46; Met1-Tyr239).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5721
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.