Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA2G4/EBP1 Antibody (OTI1D3), FITC, Novus Biologicals™
SDP

Catalog No. NB302125 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302125 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302125 Supplier Novus Biologicals Supplier No. NBP273213F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PA2G4/EBP1 Monoclonal antibody specifically detects PA2G4/EBP1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PA2G4/EBP1
Applications Western Blot
Classification Monoclonal
Clone OTI1D3
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4
Host Species Mouse
Immunogen Full length human recombinant protein of human PA2G4 (NP_006182) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5036
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.