Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PA2G4/EBP1 Antibody (OTI1D3), HRP, Novus Biologicals™
SDP

Catalog No. NBP273213H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273213H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273213H Supplier Novus Biologicals Supplier No. NBP273213H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PA2G4/EBP1 Monoclonal specifically detects PA2G4/EBP1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PA2G4/EBP1
Applications Western Blot
Classification Monoclonal
Clone OTI1D3
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4
Gene Symbols PA2G4
Host Species Mouse
Immunogen Full length recombinant protein of human PA2G4/EBP1 (NP_006182) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5036.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.