Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAAF1 Antibody (OTI1F11), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N273214J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273214J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273214J646 Supplier Novus Biologicals Supplier No. NBP273214JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAAF1 Monoclonal specifically detects PAAF1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PAAF1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F11
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ11848, PAAF, proteasomal ATPase-associated factor 1, Protein G-16, Rpn14, WD repeat domain 71, WD repeat-containing protein 71, WDR71
Gene Symbols PAAF1
Host Species Mouse
Immunogen Full length recombinant protein of human PAAF1 (NP_079431) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80227.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.