Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Pab1p Monoclonal Antibody (1G1)

Mouse Monoclonal Antibody
$331.00 - $1884.00
Specifications
| Antigen | Pab1p |
|---|---|
| Clone | 1G1 |
| Concentration | Conc. Not Determined |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA547292
|
Invitrogen™
MA547292 |
2.5 mL |
Each for $1,884.00
|
|
|||||
|
PIMA547389
|
Invitrogen™
MA547389 |
500 μL |
Each for $552.00
|
|
|||||
PIMA547390
![]() |
Invitrogen™
MA547390 |
250 μL |
Each for $331.00
|
|
|||||
Description
Pab1p Monoclonal Antibody for Western Blot, ICC/IF, IHC, IP
The Pab1p protein was discovered in Saccharomyces cerevisia since it is abundant and binds specifically to the polyA tails of mRNA molecules. The systematic name in the Saccharomyces genome database is YER165w. It has a role in polyA length control in combination with Nab2p and may also be involved in mRNA stability and transcription. Pab1p is found in both the cytoplasm and nuclei of yeast cells and has a deduced molecular weight of 64 kDa, but runs on typical SDS-PAGE gels at about 67kDa. The yeast Pab1p protein sequence includes four RNA recognition motifs followed by a single C-terminal polyA binding domain. The closest human homolog of the yeast protein is polyadenylate-binding protein cytoplasmic 1-like (PABPC1L) which has the same domain structure, although is not well conserved in amino acid sequence. There are seven human genes encoding polyA binding proteins, four of which produce proteins with the same domain structure as yeast Pab1p and human PABPC1L.Specifications
| Pab1p | |
| Conc. Not Determined | |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| Cell culture media with 5mM sodium azide | |
| P04147 | |
| 856912 | |
| Yeast polyA binding protein preparations. | |
| Antibody |
| 1G1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Yeast, Bacteria | |
| PAB1 | |
| Poly(A) binding protein | |
| PAB1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title