Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABP Antibody (10E10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N120612A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N120612A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N120612A30 Supplier Novus Biologicals Supplier No. NB1206125AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PABP Monoclonal specifically detects PABP in Human, Bovine, Canine, Primate, Xenopus, Drosophila (Negative), Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PABP
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 100000000000
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias PAB1polyadenylate-binding protein 1, PABP, PABP-1, PABP1poly(A) binding protein, cytoplasmic 2, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 1, Poly(A)-binding protein 1, poly(A)-binding protein, cytoplasmic 2
Gene Symbols PABPC1
Host Species Mouse
Immunogen Recombinant human PABP protein. [UniprotNo. P11940]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26986.0
Target Species Human, Bovine, Canine, Primate, Xenopus species, Drosophila, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.