Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABP Antibody (10E10), HRP, Novus Biologicals™
SDP

Catalog No. NB1206125H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206125H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206125H Supplier Novus Biologicals Supplier No. NB1206125H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PABP Monoclonal specifically detects PABP in Human, Bovine, Canine, Primate, Xenopus, Drosophila (Negative), Mouse (Negative) samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PABP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 100000000000
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS with No Preservative
Gene Alias PAB1polyadenylate-binding protein 1, PABP, PABP-1, PABP1poly(A) binding protein, cytoplasmic 2, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 1, Poly(A)-binding protein 1, poly(A)-binding protein, cytoplasmic 2
Gene Symbols PABPC1
Host Species Mouse
Immunogen Recombinant human PABP protein. [Uniprot# P11940]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26986
Target Species Human, Bovine, Canine, Primate, Xenopus species, Drosophila (Negative), Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.