Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABP Mouse anti-Yeast, Clone: 1G1, Novus Biologicals™
SDP

Catalog No. NB160014 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
2500 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160014 2500 μg
NB160015 250 μg
NB160016 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB160014 Supplier Novus Biologicals Supplier No. NBP3179962500UL
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PABP Monoclonal antibody specifically detects PABP in Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen PABP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 1G1
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation
Formulation Supplied as concentrated hybridoma cell culture media.
Gene Alias PAB1polyadenylate-binding protein 1, PABP, PABP-1, PABP1poly(A) binding protein, cytoplasmic 2, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 1, Poly(A)-binding protein 1, poly(A)-binding protein, cytoplasmic 2
Host Species Mouse
Immunogen Yeast polyA binding protein preparations. UniProt: P04147
Purification Method Protein G purified
Quantity 2500 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26986
Target Species Yeast
Content And Storage Store at 4C short term. Store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.