Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PABPC1L Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | PABPC1L |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121717
|
Invitrogen™
PA5121717 |
20 μL |
Each for $237.00
|
|
|||||
PIPA5121716
![]() |
Invitrogen™
PA5121716 |
100 μL |
Each for $539.50
|
|
|||||
Description
PABPC1L is a protein coding gene. Gene Ontology (GO) annotations related to this gene include RNA binding.Specifications
PABPC1L | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q4VXU2 | |
80336 | |
Recombinant Poly A Binding Protein Cytoplasmic 1 Like Protein (PABPC1L), Ala187-Lys324 (Accession Q4VXU2). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
PABPC1L |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
1810053B01Rik; C20orf119; dJ1069P2.3; embryonic poly(A) binding protein; embryonic poly(A)-binding protein; EPAB; LOW QUALITY PROTEIN: polyadenylate-binding protein 1-like; PABPC1L; PABPC1L1; poly(A) binding protein cytoplasmic 1 like; poly(A) binding protein, cytoplasmic 1-like; polyadenylate-binding protein 1-like; RGD1563142 | |
PABPC1L | |
IgG | |
Antigen affinity chromatography, Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title