Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABPC3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB327054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327054 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327054 Supplier Novus Biologicals Supplier No. NBP299107F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PABPC3 Polyclonal antibody specifically detects PABPC3 in Human samples. It is validated for Western Blot

Specifications

Antigen PABPC3
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias PABP-3, PABP3polyadenylate-binding protein 3, PABPL3testis PABP, poly(A) binding protein, cytoplasmic 3, Poly(A)-binding protein 3, poly(A)-binding protein, cytoplasmic 3, poly(A)-binding protein-like 3, Testis-specific poly(A)-binding protein, testis-specific poly(A)-binding protein (PABP), testis-specific poly(A)-binding protein 3, tPABP
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PABPC3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5042
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.