Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABPC4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10074593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074593 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074593 Supplier Novus Biologicals Supplier No. NB10074593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

PABPC4 Polyclonal specifically detects PABPC4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen) (Negative), Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot.

Specifications

Antigen PABPC4
Applications Immunohistochemistry, Immunohistochemistry (Frozen) (Negative), Immunohistochemistry (Paraffin), Immunoprecipitation, Single Cell Western, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-5 μg/mL Lysate, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen (Negative), Single Cell Western 10 μg/mL
Formulation TBS, 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_003810.1
Gene Alias APP1FLJ43938, APP-1PABP-4, Inducible poly(A)-binding protein, IPABP, iPABPpoly(A)-binding protein, cytoplasmic 4 (inducible form), PABP4Activated-platelet protein 1, poly(A) binding protein, cytoplasmic 4 (inducible form), Poly(A)-binding protein 4, polyadenylate-binding protein 4
Gene Symbols PABPC4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human Poly(A)-binding protein 4 using the numbering given in entry NP_003810.1 (GeneID 8761).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8761
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.