Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABPC4 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB103833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB103833 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB103833 Supplier Novus Biologicals Supplier No. NBP297931H

Rabbit Polyclonal Antibody

PABPC4 Polyclonal specifically detects PABPC4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PABPC4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias APP1FLJ43938, APP-1PABP-4, Inducible poly(A)-binding protein, IPABP, iPABPpoly(A)-binding protein, cytoplasmic 4 (inducible form), PABP4Activated-platelet protein 1, poly(A) binding protein, cytoplasmic 4 (inducible form), Poly(A)-binding protein 4, polyadenylate-binding protein 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PABPC4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8761
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.