Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PAC 1
GREENER_CHOICE

Catalog No. p-4784237
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Activator of procaspase-3; pro-apoptotic

Procaspase-activating compound; activates procaspase-3 to produce caspase-3 (EC50 = 0.22 μM). Also activates procaspase-7 in a less efficient manner (EC50 = 4.5 μM). Pro-apoptotic; induces apoptosis in both cancerous and non-cancerous cells dependent on procaspase-3 concentration (IC50 values are 0.003 - 1.41 and 5.02 - 9.98 μM respectively).

Chemical Identifiers

CAS 315183-21-2
Molecular Formula C23H28N4O2

Specifications

Quantity 10 mg
Formula Weight 392.5
Purity >98%
Molecular Weight (g/mol) 392.50
Chemical Name or Material PAC 1
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.