Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAC2 Antibody (OTI1E8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273215G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273215G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273215G Supplier Novus Biologicals Supplier No. NBP273215G

Mouse Monoclonal Antibody

PAC2 Monoclonal specifically detects PAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PAC2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E8
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CLAST3HSPC260, HCCA3PAC-2, hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-susceptibility protein 3, HsT1707, likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3), MDS003, MGC15092, PAC2HDCMC29P, proteasome (prosome, macropain) assembly chaperone 2, proteasome assembling chaperone 2, proteasome assembly chaperone 2, TNFSF5IP1CD40 ligand-activated specific transcript 3, Tumor necrosis factor superfamily member 5-induced protein 1, tumor necrosis factor superfamily, member 5-induced protein 1, x 003 protein
Gene Symbols PSMG2
Host Species Mouse
Immunogen Full length recombinant protein of human PAC2 (NP_064617) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 56984.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.