Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACAP/ADCYAP1 Antibody - BSA Free
SDP

Catalog No. NB10062241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062241 0.5 mL
NB017449 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062241 Supplier Novus Biologicals Supplier No. NB10062241

Sheep Polyclonal Antibody

PACAP/ADCYAP1 Polyclonal specifically detects PACAP/ADCYAP1 in Human samples. It is validated for ELISA.

Specifications

Antigen PACAP/ADCYAP1
Applications ELISA
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P18509
Gene Alias adenylate cyclase activating polypeptide 1 (pituitary), PACAPMGC126852, pituitary adenylate cyclase-activating polypeptide
Gene Symbols ADCYAP1
Host Species Sheep
Immunogen Proprietary, 14 amino acid peptide sequence of PACAP, conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 116
Test Specificity Human. Not yet tested in other species.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.