Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACS1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB163484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163484 Supplier Novus Biologicals Supplier No. NBP281695AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PACS1 Polyclonal antibody specifically detects PACS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PACS1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias cytosolic sorting protein PACS-1, FLJ10209, KIAA1175, PACS-1, phosphofurin acidic cluster sorting protein 1
Host Species Rabbit
Immunogen PACS1 antibody was raised against a 17 amino acid peptide near the center of human PACS1. The immunogen is located within amino acids 430 - 480 of PACS1. Amino Acid Squence: IKSTWIKNQDDSLTETD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55690
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.