Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACS1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224496SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224496SS 0.025 mg
NBP224496 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224496SS Supplier Novus Biologicals Supplier No. NBP224496SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PACS1 Polyclonal specifically detects PACS1 in Human, Mouse, Bovine, Canine, Equine, Opossum, Primate samples. It is validated for Western Blot.

Specifications

Antigen PACS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.05-0.1 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q6VY07
Gene Alias cytosolic sorting protein PACS-1, FLJ10209, KIAA1175, PACS-1, phosphofurin acidic cluster sorting protein 1
Gene Symbols PACS1
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human PACS-1 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55690
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_e and CRA_f), horse, cow and chimpanzee, 94% homologous in mouse and rat, 89% homologous in opossum, platypus and dog and 78% homologous in chicken.
Target Species Human, Mouse, Bovine, Canine, Equine, Opossum, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.