Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACS2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341651 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341651 Supplier Novus Biologicals Supplier No. NBP281975MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PACS2 Polyclonal antibody specifically detects PACS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PACS2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ25488, KIAA0602phosphofurin acidic cluster sorting protein 1-like, PACS1L, PACS1-like protein, PACS-2, phosphofurin acidic cluster sorting protein 2
Host Species Rabbit
Immunogen PACS2 antibody was raised against a 17 amino acid peptide near the center of human PACS2. The immunogen is located within amino acids 330 - 380 of PACS2. Amino Acid Squence: ARSHKEPPSPADVPEKT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23241
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.