Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACSIN2 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB181059 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181059 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181059 Supplier Novus Biologicals Supplier No. NBP306469FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PACSIN2 Polyclonal antibody specifically detects PACSIN2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PACSIN2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias cytoplasmic phosphoprotein PACSIN2, protein kinase C and casein kinase substrate in neurons 2, protein kinase C and casein kinase substrate in neurons protein 2, SDPII, syndapin II
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PACSIN2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11252
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.