Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACSIN3 Antibody (OTI2A3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273223UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273223UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273223UV Supplier Novus Biologicals Supplier No. NBP273223UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PACSIN3 Monoclonal specifically detects PACSIN3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PACSIN3
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI2A3
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Endophilin I, FLJ17861, protein kinase C and casein kinase substrate in neurons 3, protein kinase C and casein kinase substrate in neurons protein 3, SDPIII, SH3 domain-containing protein 6511, syndapin III
Gene Symbols PACSIN3
Host Species Mouse
Immunogen Full length recombinant protein of human PACSIN3 (NP_057307) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Diabetes Research, GPCR, Neuroscience, Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 29763.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.